Real-Time PCR detection kits, Allergen
Leverancier: BIOPREMIER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
FS-0049 FS-0050 FS-0051 FS-0052 FS-0053 FS-0054 FS-0055 FS-SP0049 FS-SP0050 FS-SP0051 FS-SP0052 FS-SP0053 FS-SP0054 FS-SP0055 FS-0060 FS-0061 FS-0062 FS-0063 FS-SP0060 FS-SP0061 FS-SP0062 FS-SP0063
BIOPFS-0049EA 684 EUR
BIOPFS-0049 BIOPFS-0050 BIOPFS-0051 BIOPFS-0052 BIOPFS-0053 BIOPFS-0054 BIOPFS-0055 BIOPFS-SP0049 BIOPFS-SP0050 BIOPFS-SP0051 BIOPFS-SP0052 BIOPFS-SP0053 BIOPFS-SP0054 BIOPFS-SP0055 BIOPFS-0060 BIOPFS-0061 BIOPFS-0062 BIOPFS-0063 BIOPFS-SP0060 BIOPFS-SP0061 BIOPFS-SP0062 BIOPFS-SP0063
Real-Time PCR detection kits, Allergen
Testsystemen voor microbiologie PCR testen
These kits allow the detection of allergic substances. Ideal for controlling and monitoring the various steps of food production.