Anti-Neurokinin B Receptor Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-0166R-A680
BOSSBS-0166R-A680EA 899 EUR
BOSSBS-0166R-A680
Anti-Neurokinin B Receptor Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antilichamen
This is a receptor for the tachykinin neuropeptide neuromedin-K (neurokinin B). It is associated with G proteins that activate a phosphatidylinositol-calcium second messenger system. The rank order of affinity of this receptor to tachykinins is: neuromedin-K > substance K > substance P.

Type: Primary
Antigen: Neurokinin B receptor
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 151-250/440
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Specificaties

Environmentally Preferable
Type antilichaam Primair,Neurokinin B receptor
Antigeen synoniemen Neurokinin B receptor|Tachykinin receptor 3|Neuromedin-K receptor|TAC3RL|TACR3|NKR|NK3R|NK-3R|HH11|NK-3 receptor|TAC3R
Type antilichaam Primair,Neurokinin B receptor
Clonality Polyklonaal
Conjugatie Alexa Fluor® 680
Reactivity Human,Rat,Mouse
Host Rabbit
Gen-ID 6870
Isotype IgG
ImmunoChemie Yes
ImmunoFluorescentie Yes
Flowcytometrie Yes
Modification Unmodified
Epitoop 151-250/440
Immunogeen KLH conjugated synthetic peptide derived from human NKR
Formulier Liquid
Moleculair gewicht 52 kDa
Purification Purified by Protein A
Concentratie 1 µg/µl
Opslagbuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Opslagtemperatuur Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Shipping temperature 4 °C
Tested applications ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR