Anti-LDLR Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-0705R-A350
BOSSBS-0705R-A350EA 901 EUR
BOSSBS-0705R-A350
Anti-LDLR Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antilichamen
Binds LDL, the major cholesterol-carrying lipoprotein of plasma, and transports it into cells by endocytosis. In order to be internalized, the receptor-ligand complexes must first cluster into clathrin-coated pits. In case of HIV-1 infection, functions as a receptor for extracellular Tat in neurons, mediating its internalization in uninfected cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LDLR
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Other Species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR