Anti-GFP Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Leverancier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-0890R-A555
BOSSBS-0890R-A555EA 893 EUR
BOSSBS-0890R-A555
Anti-GFP Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antilichamen
Green fluorescence protein (GFP) is a 27 kDa protein derived from the jellyfish Aequorea victoria, which emits green light (emission peak at a wavelenth of 509 nm) when excited by blue light (excitation peak at a wavelenth of 395 nm). Green Fluorescent Protein (GFP) has become an invaluable tool in cell biology research, since its intrinsic fluorescence can be visualized in living cells. GFP fluorescence is stable under fixation conditions and suitable for a variety of applications. GFP has been widely used as a reporter for gene expression, enabling researchers to visualize and localize GFP-More Product Information