Anti-GRIA1 Rabbit Polyclonal Antibody (Cy5®)
Leverancier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-10042R-CY5
BOSSBS-10042R-CY5EA 909 EUR
BOSSBS-10042R-CY5
Anti-GRIA1 Rabbit Polyclonal Antibody (Cy5®)
Antilichamen
Ionotropic glutamate receptor. L-glutamate acts as an excitatory neurotransmitter at many synapses in the central nervous system. Binding of the excitatory neurotransmitter L-glutamate induces a conformation change, leading to the opening of the cation channel, and thereby converts the chemical signal to an electrical impulse. The receptor then desensitizes rapidly and enters a transient inactive state, characterized by the presence of bound agonist. In the presence of CACNG4 or CACNG7 or CACNG8, shows resensitization which is characterized by a delayed accumulation of current flux uponMore Product Information