Anti-TNFAIP8L2/TIPE2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Leverancier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-11035R-A750
BOSSBS-11035R-A750EA 890 EUR
BOSSBS-11035R-A750
Anti-TNFAIP8L2/TIPE2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antilichamen
TNF Alpha-IP 8L2 (tumour necrosis factor, alpha-induced protein 8-like 2), also known as TIPE2, is a 184 amino acid protein that shares 94% identity with its mouse counterpart and belongs to the TNFAIP8 family. Expressed in spleen, thymus, small intestine and lymph node with lower levels present in colon, lung and skin, TNF Alpha-IP 8L2 plays a role in maintaining immune homeostasis, specifically by acting as a negative regulator of both innate and adaptive immunity. In addition, TNF IP 8L2 functions as a negative regulator of T-cell receptor function and is thought to promote Fas-inducedMore Product Information