Anti-CALNB1 Rabbit Polyclonal Antibody (Cy3®)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-11246R-CY3
BOSSBS-11246R-CY3EA 900 EUR
BOSSBS-11246R-CY3
Anti-CALNB1 Rabbit Polyclonal Antibody (Cy3®)
Antilichamen
In eukaryotes, the phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions including division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the protein phosphatases. In general, the protein phosphatase (PP) holoenzyme is a trimeric complex composed of a regulatory subunit, a variable subunit and a catalytic subunit. Four major families of protein phosphatase catalytic subunit have been identified, designated PP1, PP2A, PP2B and PP2C. AnMore Product Information
Order Now

Specificaties

antigeen symbool CALNB1
Type antilichaam Primair,Calcineurin B
Environmentally Preferable
Antigeen synoniemen PPP3R1 protein phosphatase 3 formerly 2B|regulatory subunit B|CNB|Ppp3r1|19kDa|alpha isoform Protein phosphatase 2B regulatory subunit 1|protein phosphatase3 formerly2B|CANB1_HUMAN|CNB1|Cna2|Protein phosphatase 2B regulatory subunit B alpha|calcineurin B|CALNB1|OTTHUMP00000201960|alpha isoform|Protein phosphatase 3 regulatory subunit B alpha|Protein phosphatase 3 regulatory subunit B alpha isoform 1.|OTTHUMP00000201961|Calcineurin subunit B type 1|protein phosphatase 3 formerly 2B|type I 19kDa|type I|alpha isoform calcineurin B
Type antilichaam Primair,Calcineurin B
Clonality Polyklonaal
Conjugatie Cy3®
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype IgG
ImmunoChemie Yes
Formulier liquid
Purification Purified by Protein A
Concentratie 1 μg/μl
Kruisadsorptie No
Opslagbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Opslagtemperatuur Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR