Anti-FAM126A Rabbit Polyclonal Antibody
Leverancier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-11554R
BOSSBS-11554REA 725 EUR
BOSSBS-11554R
Anti-FAM126A Rabbit Polyclonal Antibody
Antilichamen
Hyccin is a 521 amino acid cytoplasmic protein that is widely expressed with highest levels found in heart, brain, placenta, spleen and testis. Belonging to the FAM126 family, hyccin may play a role in the ∫-catenin/Lef signaling pathway. Hyccin is likely involved in the process of myelination of the central and peripheral nervous system. Defects in the gene encoding hyccin are the cause of leukodystrophy hypomyelinating type 5 (HLD5), which is characterized by congenital cataract, progressive neurologic impairment and diffuse myelin deficiency. Individuals affected by HLD5 experienceMore Product Information