Anti-SOX13 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-11586R-A350
BOSSBS-11586R-A350EA 893 EUR
BOSSBS-11586R-A350
Anti-SOX13 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antilichamen
Sox genes comprise a family of genes that are related to the mammalian sex determining gene SRY. These genes similarly contain sequences that encode for the HMG-box domain, which is responsible for the sequence-specific DNA-binding activity. Sox genes encode putative transcriptional regulators implicated in the decision of cell fates during development and the control of diverse developmental processes. The highly complex group of Sox genes cluster at least 40 different loci that rapidly diverged in various animal lineages. At present, 30 Sox genes have been identified. Members of thisMore Product Information
Order Now

Specificaties

antigeen symbool SOX13
Environmentally Preferable
Type antilichaam Primair,SRY (sex determining region Y)-box 13
Antigeen synoniemen MGC117216|SOX 13|ICA 12|sox13|SOX13 protein|Transcription factor Sox-13|Transcription factor SOX 13|ICA12|SRY related HMG box gene 13|Islet cell antigen 12|Type 1 diabetes autoantigen|SRY Sex determining region Y-box 13|SOX13_HUMAN|SRY box 13|SRY sex determining region Y box 13|SOX 13 protein|SRY box containing gene 13|Islet cell 12|Type 1 diabetes autoantigen ICA 12|Type 1 diabetes autoantigen ICA12.
Type antilichaam Primair,SRY (sex determining region Y)-box 13
Clonality Polyklonaal
Conjugatie Alexa Fluor® 350
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype IgG
ImmunoChemie Yes
Formulier liquid
Purification Purified by Protein A
Concentratie 1 μg/μl
Kruisadsorptie No
Opslagbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Opslagtemperatuur Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR