Anti-SCGN Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-11754R-A647
BOSSBS-11754R-A647EA 893 EUR
BOSSBS-11754R-A647
Anti-SCGN Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antilichamen
SCGN is a 276 amino acid cytoplasmic protein that contains six EF-hand domains and is related to the calicium-binding proteins Calretinin and Calbindin D28K. Expressed in a variety of tissues including stomach, thyroid, colon, brain and neuroendocrine cells, SCGN is thought to be involved in cell proliferation and KCl (potassium chloride)-mediated calcium flux events. Through its interaction with KCl and its subsequent ability to modulate calcium storage pools within the cell, SCGN may function to negatively control growth and differentiation rates and, thus, indirectly inhibit cellMore Product Information
Order Now

Specificaties

antigeen symbool SCGN
Type antilichaam Primair,Secretagogin, EF-hand Calcium binding Protein
Environmentally Preferable
Antigeen synoniemen DJ501N12.8|Secretagogin|SEGN|SECRET|SEGN_HUMAN|Secretagogin EF hand Calcium binding protein|Calbindin like|Setagin.|Scgn|CALBL
Type antilichaam Primair,Secretagogin, EF-hand Calcium binding Protein
Clonality Polyklonaal
Conjugatie Alexa Fluor® 647
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype IgG
ImmunoChemie Yes
Formulier liquid
Purification Purified by Protein A
Concentratie 1 μg/μl
Kruisadsorptie No
Opslagbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Opslagtemperatuur Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR