Anti-LRRTM4 Rabbit Polyclonal Antibody (Cy5®)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-11878R-CY5
BOSSBS-11878R-CY5EA 900 EUR
BOSSBS-11878R-CY5
Anti-LRRTM4 Rabbit Polyclonal Antibody (Cy5®)
Antilichamen
The leucine-rich (LRR) repeat is a 20-30 amino acid motif that forms a hydrophobic å/∫ horseshoe fold, allowing it to accommodate several leucine residues within a tightly packed core. All LRR repeats contain a variable segment and a highly conserved segment, the latter of which accounts for 11 or 12 residues of the entire LRR motif. The LRRTM protein family plays a role in the regulation of various cellular events during nervous system development. Localizing predominantly to the nervous system, LRRTM family members are known to exhibit synaptogenic activity. LRRTM4 (leucine-rich repeatMore Product Information
Order Now

Specificaties

antigeen symbool LRRTM4
Environmentally Preferable
Type antilichaam Primair,Leucine Rich Repeat Transmembrane Neuronal 4
Type antilichaam Primair,Leucine Rich Repeat Transmembrane Neuronal 4
Clonality Polyklonaal
Conjugatie Cy5®
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype IgG
ImmunoChemie Yes
Flowcytometrie Yes
Formulier liquid
Purification Purified by Protein A
Concentratie 1 μg/μl
Kruisadsorptie No
Opslagbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Opslagtemperatuur Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR