Anti-LGI3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-11879R-FITC
BOSSBS-11879R-FITCEA 900 EUR
BOSSBS-11879R-FITC
Anti-LGI3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antilichamen
The leucine-rich (LRR) repeat is a 20-30 amino acid motif that forms a hydrophobic å/∫ horseshoe fold, allowing it to accommodate several leucine residues within a tightly packed core. All LRR repeats contain a variable segment and a highly conserved segment, the latter of which accounts for 11 or 12 residues of the entire LRR motif. Leucine-rich glioma-inactivated protein 3 (LGI3), also known as LGI1-like protein 4 (LGIL4) or leucine-rich repeat LGI family member 3, is a 548 amino acid secretory protein. LGI3 contains five LRR repeats and seven EAR repeats. Widely expressed, with highestMore Product Information
Order Now

Specificaties

antigeen symbool LGI3
Type antilichaam Primair,Leucine-rich Repeat LGI family member 3
Environmentally Preferable
Antigeen synoniemen LGIL4|LGI3|UNQ8190/PRO23199|Leucine-rich repeat LGI family member 3|LGI1-like protein 4
Type antilichaam Primair,Leucine-rich Repeat LGI family member 3
Clonality Polyklonaal
Conjugatie FITC (Fluoresceïne-isothiocyanaat)
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype IgG
ImmunoChemie Yes
Formulier liquid
Purification Purified by Protein A
Concentratie 1 μg/μl
Kruisadsorptie No
Opslagbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Opslagtemperatuur Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR