Anti-SIM2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Leverancier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-11951R-A488
BOSSBS-11951R-A488EA 893 EUR
BOSSBS-11951R-A488
Anti-SIM2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antilichamen
The Per-Arnt-Sim (PAS) domain was identified as a 270 amino acid motif that mediates associations between various PAS family transcription factors. Several PAS domain family members have been identified including AhR, Arnt 1, and single-minded proteins (SIM1 and SIM2). The aromatic (aryl) hydrocarbon receptor, AhR, is a ligand dependent transcription factor that interacts with specific DNA sequences termed xenobiotic responsive elements (XREs) to activate several genes including CYP1A1, glutathione S-transferase Ya subunit and DT-diaphorase. The Ah receptor nuclear translocator protein 1 (More Product Information