Anti-SIM2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-11951R-A680
BOSSBS-11951R-A680EA 890 EUR
BOSSBS-11951R-A680
Anti-SIM2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antilichamen
The Per-Arnt-Sim (PAS) domain was identified as a 270 amino acid motif that mediates associations between various PAS family transcription factors. Several PAS domain family members have been identified including AhR, Arnt 1, and single-minded proteins (SIM1 and SIM2). The aromatic (aryl) hydrocarbon receptor, AhR, is a ligand dependent transcription factor that interacts with specific DNA sequences termed xenobiotic responsive elements (XREs) to activate several genes including CYP1A1, glutathione S-transferase Ya subunit and DT-diaphorase. The Ah receptor nuclear translocator protein 1 (More Product Information
Order Now

Specificaties

Environmentally Preferable
Type antilichaam Primair,SIM2
Antigeen synoniemen SIM|SIM 2|Transcription factor SIM2|Sim2|Single-minded homolog 2|MGC119447|Class E basic helix loop helix protein 15|SIM2_HUMAN|Single minded homolog 2 Drosophila|Class E basic helix-loop-helix protein 15|Single minded homolog 2|bHLHe15
Type antilichaam Primair,SIM2
Clonality Polyklonaal
Conjugatie Alexa Fluor® 680
Reactivity
Host Rabbit
Isotype IgG
ImmunoChemie Yes
ImmunoFluorescentie Yes
Modification Unmodified
Epitoop 321-430/667
Immunogeen KLH conjugated synthetic peptide derived from human SIM2
Formulier Liquid
Moleculair gewicht 73 kDa
Purification Purified by Protein A
Concentratie 1 µg/µl
Opslagbuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Opslagtemperatuur Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Shipping temperature 4 °C
Tested applications ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR