Anti-PPP2R5D Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-11992R-HRP
BOSSBS-11992R-HRPEA 900 EUR
BOSSBS-11992R-HRP
Anti-PPP2R5D Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antilichamen
In eukaryotes, the phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions, including division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the protein phosphatases. In general, the protein phosphatase (PP) holoenzyme is a trimeric complex composed of a regulatory subunit, a variable subunit, and a catalytic subunit. Four major families of protein phosphatase catalytic subunits have been identified, designated PP1, PP2A, PP2B (calcineurin) andMore Product Information
Order Now

Specificaties

antigeen symbool PPP2R5D
Type antilichaam Primair,Protein Phosphatase 2, Regulatory Subunit 5D
Environmentally Preferable
Antigeen synoniemen B subunit|56 kDa regulatory subunit|B56 delta isoform|B56D|protein phosphatase 2A|PR61 delta isoform|B' delta isoform|delta isoform|delta isoform of regulatory subunit B56|PPP2R5D|regulatory subunit B'|Serine/threonine protein phosphatase 2A|serine/threonine-protein phosphatase 2A 56 kDa re|regulatory subunit B (B56)|MGC2134|PP2A|MGC8949|R5 delta isoform|protein phosphatase 2
Type antilichaam Primair,Protein Phosphatase 2, Regulatory Subunit 5D
Clonality Polyklonaal
Conjugatie HRP (Horseradish Peroxidase)
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoChemie Yes
Formulier liquid
Purification Purified by Protein A
Concentratie 1 μg/μl
Kruisadsorptie No
Opslagbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
Opslagtemperatuur Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR