Anti-GRIK2 Rabbit Polyclonal Antibody (Cy7®)
Leverancier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-12006R-CY7
BOSSBS-12006R-CY7EA 893 EUR
BOSSBS-12006R-CY7
Anti-GRIK2 Rabbit Polyclonal Antibody (Cy7®)
Antilichamen
Glutamate receptors mediate most excitatory neurotransmission in the brain and play an important role in neural plasticity, neural development and neurodegeneration. Ionotropic glutamate receptors are categorized into NMDA receptors and kainate/AMPA receptors, both of which contain glutamate-gated, cation-specific ion channels. Kainate/AMPA receptors are co-localized with NMDA receptors in many synapses and consist of seven structurally related subunits designated GluR-1 to -7. The kainate/AMPA receptors are primarily responsible for the fast excitatory neuro-transmission by glutamate,More Product Information