Anti-GABRB2/GABA A Receptor beta 2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-12065R-A750
BOSSBS-12065R-A750EA 890 EUR
BOSSBS-12065R-A750
Anti-GABRB2/GABA A Receptor beta 2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antilichamen
GAD-65 and GAD-67, glutamate decarboxylases, function to catalyse the production of GABA (gamma-aminobutyric acid). In the central nervous system GABA functions as the main inhibitory transmitter by increasing a Cl- conductance that inhibits neuronal firing. GABA has been shown to activate both ionotropic (GABAA) and metabotropic (GABAB) receptors as well as a third class of receptors called GABAC. Both GABAA and GABAC are ligand-gated ion channels, however, they are structurally and functionally distinct.

Type: Primary
Antigen: GABRB2/GABA A Receptor beta 2
Clonality: Polyclonal
Clone:
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR