Anti-LRRC39 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Leverancier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-12312R-A350
BOSSBS-12312R-A350EA 893 EUR
BOSSBS-12312R-A350
Anti-LRRC39 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antilichamen
The leucine-rich (LRR) repeat is a 20-30 amino acid motif that forms a hydrophobic å/∫ horseshoe fold, allowing it to accommodate several leucine residues within a tightly packed core. All LRR repeats contain a variable segment and a highly conserved segment, the latter of which accounts for 11 or 12 residues of the entire LRR motif. The primary function of these motifs is to provide a versatile structural framework to mediate the formation of protein-protein interactions. LRRs are present in a variety of proteins with diverse structure and function, including innate immunity and nervousMore Product Information