Anti-STK33 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Leverancier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-12373R-A488
BOSSBS-12373R-A488EA 893 EUR
BOSSBS-12373R-A488
Anti-STK33 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antilichamen
The phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions in eukaryotes, including cell division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the serine/threonine (Ser/Thr) protein kinases. STK33 (serine/threonine kinase 33) is a 514 amino acid protein that belongs to the CaMK (calcium/calmodulin dependent kinase) subfamily of structurally related serine/threonine kinases. Widely expressed at low levels with predominant expression in testis,More Product Information