Anti-EFNB1 Rabbit Polyclonal Antibody (Cy7®)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-13085R-CY7
BOSSBS-13085R-CY7EA 1350 EUR
BOSSBS-13085R-CY7
Anti-EFNB1 Rabbit Polyclonal Antibody (Cy7®)
Antilichamen
Ephrin B proteins are thought to play key roles in cellular functions as diverse as neuronal migration and blood vessel development. Ephrin B molecules expressed at the membrane surface bind to the Ephrin B family receptors on target cells during cell to cell contact. This interaction leads to cell signaling in the target cell but also generates a reverse signal in the cell expressing Ephrin B on its surface. This reverse signaling event is thought to be critical for vessel maturation and neuronal development. Importantly, tyrosine phosphorylation of Ephrin B is thought to be a criticalMore Product Information
Order Now

Specificaties

antigeen symbool EFNB1
Environmentally Preferable
Type antilichaam Primair,Ephrin B1
Antigeen synoniemen Elk-L|CFNS|EFB1|LERK2|EFL3|CFND|EPLG2
Type antilichaam Primair,Ephrin B1
Clonality Polyklonaal
Conjugatie Cy7®
Reactivity Human,Mouse
Host Rabbit
Gen-ID 1947
Isotype IgG
ImmunoChemie Yes
Modification phospho-Ser287
Formulier liquid
Purification Purified by Protein A
Concentratie 1 μg/μl
Kruisadsorptie No
Opslagbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Opslagtemperatuur Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR