Anti-GALM Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Leverancier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-13268R-A488
BOSSBS-13268R-A488EA 893 EUR
BOSSBS-13268R-A488
Anti-GALM Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antilichamen
Galactose Mutarotase is a member of the aldose epimerase family and is involved in hexose metabolism. Through its catalytic activity, Galactose Mutarotase converts beta-aldose to alpha-aldose on several sugars, including D-glucose, L-arabinose and D-xylose. Found in the cytoplasm of most cells, Galactose Mutarotase plays a key role in galactose metabolism by catalyzing the conversion of beta-D-galactose to alpha-D-galactose. The enzyme contains two residues, Glu 304 and His 170, that are critical for catalysis, as well as His 96 and Asp 243, which are important for proper substrateMore Product Information