Anti-GALNT10 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Leverancier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-13270R-HRP
BOSSBS-13270R-HRPEA 900 EUR
BOSSBS-13270R-HRP
Anti-GALNT10 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antilichamen
The UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase (GalNAc-T) family of enzymes are substrate-specific proteins that catalyze the transfer of GalNAc (N-acetylgalactosaminyl) to serine and threonine residues of various proteins, thereby initiating mucin-type O-linked glycosylation in the Golgi apparatus. GalNAc-T10 (Polypeptide N-acetylgalactosaminyltransferase 10), also known as UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 10, is a 603 amino acid single-pass type II membrane protein that prefers Muc5Ac and EA2 peptide substrates. The N-terminalMore Product Information