Anti-GAPDHS Rabbit Polyclonal Antibody (Cy3®)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-13282R-CY3
BOSSBS-13282R-CY3EA 909 EUR
BOSSBS-13282R-CY3
Anti-GAPDHS Rabbit Polyclonal Antibody (Cy3®)
Antilichamen
May play an important role in regulating the switch between different pathways for energy production during spermiogenesis and in the spermatozoon. Required for sperm motility and male fertility (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GAPDHS/GAPDH2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR