Anti-GBA3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-13298R-A555
BOSSBS-13298R-A555EA 893 EUR
BOSSBS-13298R-A555
Anti-GBA3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antilichamen
CBG is a monomeric enzyme involved in the absorption and metabolism of flavonoid glucosides. CBG is found predominately in the liver, but is also located in tissues such as spleen, small intestine and kidney. Through its catalytic activity, CBG is able to hydrolyze a variety of glycosides including phytoestrogens, cyanogens, and flavonols. Although its catalytic activity extends to many dietary flavonoids, CBG has increased specificity for hydrophobic aglycones such as beta-D-glucoside and beta-D-galactoside. Hydrolysis is inhibited by sodium taurocholate and glucosyl-sphingosine, both ofMore Product Information
Order Now

Specificaties

antigeen symbool GBA3
Environmentally Preferable
Type antilichaam Primair,Glucosidase, Beta, Acid 3 (Gene/Pseudogene)
Antigeen synoniemen CBGL1|KLRP|GLUC|CBG
Type antilichaam Primair,Glucosidase, Beta, Acid 3 (Gene/Pseudogene)
Clonality Polyklonaal
Conjugatie Alexa Fluor® 555
Reactivity Human,Rat
Host Rabbit
Gen-ID 57733
Isotype IgG
ImmunoChemie Yes
Formulier liquid
Purification Purified by Protein A
Concentratie 1 μg/μl
Kruisadsorptie No
Opslagbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Opslagtemperatuur Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR