Anti-Gcn1l1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-13315R-A680
BOSSBS-13315R-A680EA 890 EUR
BOSSBS-13315R-A680
Anti-Gcn1l1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antilichamen
GCN1L1 is a 2,671 amino acid protein that is ubiquitously expressed and belongs to the GCN1 family. Functioning as a translation activator, GCN1L1 interacts with IMPACT to regulproteinate GCN2 kinase activity. GCN1L1 contains 24 HEAT repeats and is encoded by a gene that maps to human chromosome 12q24.23. Chromosome 12 encodes over 1,100 genes and comprises approximately 4.5% of the human genome. Chromosome 12 is associated with a variety of diseases and afflictions, including hypochondrogenesis, achondrogenesis, Kniest dysplasia, Noonan syndrome and trisomy 12p, which causes facialMore Product Information
Order Now

Specificaties

Environmentally Preferable
Type antilichaam Primair,Gcn1l1
Antigeen synoniemen GCN1L|GCN1 general control of amino acid synthesis 1 like 1|GCN1L_HUMAN|GCN1 general control of amino acid synthesis 1 like 1 yeast|GCN1 general control of amino acid synthesis yeast homolog like|GCN1 like protein 1|GCN1 general control of amino acid synthesis 1 yeast like 1|HsGCN1|GCN1
Type antilichaam Primair,Gcn1l1
Clonality Polyklonaal
Conjugatie Alexa Fluor® 680
Reactivity
Host Rabbit
Gen-ID 10985
Isotype IgG
Western blot Yes
ImmunoChemie Yes
ImmunoFluorescentie Yes
Modification Unmodified
Epitoop 751-850/2671
Immunogeen KLH conjugated synthetic peptide derived from human Gcn1l1
Formulier Liquid
Moleculair gewicht 293 kDa
Purification Purified by Protein A
Concentratie 1 µg/µl
Opslagbuffer Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
Opslagtemperatuur Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Shipping temperature 4 °C
Tested applications ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR