Anti-Glucoamylase Rabbit Polyclonal Antibody

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-13383R
BOSSBS-13383REA 725 EUR
BOSSBS-13383R
Anti-Glucoamylase Rabbit Polyclonal Antibody
Antilichamen
Glucoamylase is an important industrial enzyme used in saccharification steps in both starch enzymatic conversion and in alcohol production. The catalytic domain degrades oligosaccharides from the non reducing end, releasing glucose, and the starch domain binds the enzyme to raw starch and to the cell wall.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: Glucoamylase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Other Species
Order Now

Specificaties

Environmentally Preferable
Type antilichaam Primair,Glucoamylase
Antigeen synoniemen Glucan 1,4|AMYG_ASPAW|Glucoamylase 3|Glucan 14 alpha glucosidase|Amyloglucosidase|Glucoamylase|Glucan 1 4 alpha glucosidase|Gluc 2|1,4-alpha-D-glucan glucohydrolase|1 4 alpha D glucan glucohydrolase|Gluc 1|Gluc 3|Glucoamylase 2|Glucoamylase 1
Type antilichaam Primair,Glucoamylase
Clonality Polyklonaal
Conjugatie Onconjugeerd
Reactivity Multiple
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoChemie Yes
Formulier liquid
Purification Purified by Protein A.
Concentratie 1 ug/ul
Opslagbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
Shipping temperature 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR