Anti-C7ORF69 Rabbit Polyclonal Antibody (Cy5)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-15281R-CY5
BOSSBS-15281R-CY5EA 900 EUR
BOSSBS-15281R-CY5
Anti-C7ORF69 Rabbit Polyclonal Antibody (Cy5)
Antilichamen
C7orf69.

Type: Primary
Antigen: C7orf69
Clonality: Polyclonal
Clone:
Conjugation: Cy5
Public Immunogen Range: 31-122/122
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR