Anti-H2A1J Rabbit Polyclonal Antibody (Cy3)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-15399R-CY3
BOSSBS-15399R-CY3EA 900 EUR
BOSSBS-15399R-CY3
Anti-H2A1J Rabbit Polyclonal Antibody (Cy3)
Antilichamen
H2A1J.

Type: Primary
Antigen: H2A1J
Clonality: Polyclonal
Clone:
Conjugation: Cy3
Public Immunogen Range: 1-100/128
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR