Anti-HBQ1 Rabbit Polyclonal Antibody (ALEXA FLUOR® 488)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-15419R-A488
BOSSBS-15419R-A488EA 893 EUR
BOSSBS-15419R-A488
Anti-HBQ1 Rabbit Polyclonal Antibody (ALEXA FLUOR® 488)
Antilichamen
Theta-globin mRNA is found in human fetal erythroid tissue but not in adult erythroid or other nonerythroid tissue. The theta-1 gene may be expressed very early in embryonic life, perhaps sometime before 5 weeks. Theta-1 is a member of the human alpha-globin gene cluster that involves five functional genes and two pseudogenes. The order of genes is: 5' - seta - pseudoseta - mu - pseudoalpha-2 -pseudoalpha-1 - alpha-2 - alpha-1 - theta-1 - 3'. Research supports a transcriptionally active role for the gene and a functional role for the peptide in specific cells, possibly those of earlyMore Product Information
Order Now

Specificaties

Type antilichaam Primair,HBQ1
Environmentally Preferable
Antigeen synoniemen Theta 1 globin|Hemoglobin subunit theta-1|Hemoglobin theta 1|HBAT_HUMAN|Hemoglobin theta 1 chain|Theta-1-globin|Hemoglobin theta-1 chain|Hemoglobin subunit theta 1|HBQ1
Type antilichaam Primair,HBQ1
Clonality Polyklonaal
Conjugatie Alexa Fluor® 488
Reactivity Human
Host Rabbit
Gen-ID 3049
Isotype IgG
Western blot Yes
ImmunoChemie Yes
ImmunoFluorescentie Yes
Modification Unmodified
Epitoop 1-100/142
Immunogeen KLH conjugated synthetic peptide derived from human HBQ1
Formulier Liquid
Moleculair gewicht 15 kDa
Purification Purified by Protein A
Concentratie 1 µg/µl
Opslagbuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Opslagtemperatuur Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Shipping temperature 4 °C
Tested applications ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR