Anti-DRD3 Rabbit Polyclonal Antibody

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-1743R
BOSSBS-1743REA 752 EUR
BOSSBS-1743R
Anti-DRD3 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: DRD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR