Anti-AURKB Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Leverancier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-2445R-A647
BOSSBS-2445R-A647EA 901 EUR
BOSSBS-2445R-A647
Anti-AURKB Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antilichamen
Serine/threonine-protein kinase component of the chromosomal passenger complex (CPC), a complex that acts as a key regulator of mitosis. The CPC complex has essential functions at the centromere in ensuring correct chromosome alignment and segregation and is required for chromatin-induced microtubule stabilization and spindle assembly. Involved in the bipolar attachment of spindle microtubules to kinetochores and is a key regulator for the onset of cytokinesis during mitosis. Required for central/midzone spindle assembly and cleavage furrow formation. Key component of the cytokinesisMore Product Information