Anti-ICOS Rabbit Polyclonal Antibody (Cy5®)
Leverancier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-2583R-CY5
BOSSBS-2583R-CY5EA 900 EUR
BOSSBS-2583R-CY5
Anti-ICOS Rabbit Polyclonal Antibody (Cy5®)
Antilichamen
Enhances all basic T-cell responses to a foreign antigen, namely proliferation, secretion of lymphokines, up-regulation of molecules that mediate cell-cell interaction, and effective help for antibody secretion by B-cells. Essential both for efficient interaction between T and B-cells and for normal antibody responses to T-cell dependent antigens. Does not up-regulate the production of interleukin-2, but superinduces the synthesis of interleukin-10. Prevents the apoptosis of pre-activated T-cells. Plays a critical role in CD40-mediated class switching of immunoglobin isotypes (By similarity).More Product Information