Anti-SPHK2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-2653R-A680
BOSSBS-2653R-A680EA 899 EUR
BOSSBS-2653R-A680
Anti-SPHK2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antilichamen
Catalyses the phosphorylation of sphingosine to form sphingosine 1-phosphate (SPP), a lipid mediator with both intra- and extracellular functions. Also acts on D-erythro-dihydrosphingosine, D-erythro-sphingosine and L-threo-dihydrosphingosine. Binds phosphoinositides.

Type: Primary
Antigen: SPHK2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/654
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR