Anti-S1PR4 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-2883R-A647
BOSSBS-2883R-A647EA 893 EUR
BOSSBS-2883R-A647
Anti-S1PR4 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antilichamen
This protein belongs to a G protein coupled heptahelical receptor subfamily named Endothelial Cell Differentiation Genes (EDG)that act as receptors for biologically active lysophospholipids. This group consists of two receptor subgroups specific for S1P and LPA respectively. EDG6 is the receptor for lysophospholipid sphingosine 1 phosphate (S1P). S1P elicits diverse physiological effect on most types of cells and tissues. EDG6 may be involved in cell migration processes that are specific for lymphocytes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: S1PR4
Clonality:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR