Anti-CEBPB Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Leverancier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-3062R-HRP
BOSSBS-3062R-HRPEA 1350 EUR
BOSSBS-3062R-HRP
Anti-CEBPB Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antilichamen
The protein encoded by this intronless gene is a bZIP transcription factor which can bind as a homodimer to certain DNA regulatory regions. It can also form heterodimers with the related proteins CEBP alpha, CEBP delta, and CEBP gamma. The encoded protein is important in the regulation of genes involved in immune and inflammatory responses and has been shown to bind to the IL1 response element in the IL6 gene, as well as to regulatory regions of several acute-phase and cytokine genes. In addition, the encoded protein can bind the promoter and upstream element and stimulate the expression ofMore Product Information