Anti-TOPO2A Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Leverancier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-3451R-A488
BOSSBS-3451R-A488EA 921 EUR
BOSSBS-3451R-A488
Anti-TOPO2A Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antilichamen
This gene encodes a DNA topoisomerase, an enzyme that controls and alters the topologic states of DNA during transcription. This nuclear enzyme is involved in processes such as chromosome condensation, chromatid separation, and the relief of torsional stress that occurs during DNA transcription and replication. It catalyzes the transient breaking and rejoining of two strands of duplex DNA which allows the strands to pass through one another, thus altering the topology of DNA. Two forms of this enzyme exist as likely products of a gene duplication event. The gene encoding this form, alpha, isMore Product Information