Anti-VASP Rabbit Polyclonal Antibody (Cy7®)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-3603R-CY7
BOSSBS-3603R-CY7EA 900 EUR
BOSSBS-3603R-CY7
Anti-VASP Rabbit Polyclonal Antibody (Cy7®)
Antilichamen
Vasodilator-stimulated phosphoprotein (VASP) is a member of the Ena-VASP protein family. Ena-VASP family members contain an EHV1 N-terminal domain that binds proteins containing E/DFPPPPXD/E motifs and targets Ena-VASP proteins to focal adhesions. In the mid-region of the protein, family members have a proline-rich domain that binds SH3 and WW domain-containing proteins. Their C-terminal EVH2 domain mediates tetramerization and binds both G and F actin. VASP is associated with filamentous actin formation and likely plays a widespread role in cell adhesion and motility. VASP may also beMore Product Information
Order Now

Specificaties

antigeen symbool VASP
Environmentally Preferable
Type antilichaam Primair,Vasodilator-stimulated phosphoprotein
Type antilichaam Primair,Vasodilator-stimulated phosphoprotein
Clonality Polyklonaal
Conjugatie Cy7®
Reactivity Human,Rat,Mouse
Host Rabbit
Gen-ID 7408
Isotype IgG
ImmunoChemie Yes
Formulier liquid
Purification Purified by Protein A
Concentratie 1 μg/μl
Kruisadsorptie No
Opslagbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Opslagtemperatuur Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR