Anti-NCF/NCF4/p40phox Rabbit Polyclonal Antibody (Cy5®)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-3616R-CY5
BOSSBS-3616R-CY5EA 900 EUR
BOSSBS-3616R-CY5
Anti-NCF/NCF4/p40phox Rabbit Polyclonal Antibody (Cy5®)
Antilichamen
NCF4 is a cytosolic oxidase associated protein. It is a component of the nicotinamide adenine dinucleotide phosphate oxidase, which mediates down regulation of NADPH oxidase through interactions with its SH3 domain. NCF4 is associated with p67 phox but is absent in patients with chronic granulomatous disease who lack p67 phox.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NCF/NCF4/p40phox
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Specificaties

antigeen symbool NCF/NCF4/p40phox
Environmentally Preferable
Type antilichaam Primair
Clonality Polyklonaal
Conjugatie Cy5®
Reactivity Human,Rat,Mouse
Host Rabbit
Gen-ID 4689
Isotype IgG
ImmunoChemie Yes
Formulier liquid
Purification Purified by Protein A
Concentratie 1 μg/μl
Kruisadsorptie No
Opslagbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Opslagtemperatuur Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR