Anti-SLC44A1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-3667R-A647
BOSSBS-3667R-A647EA 901 EUR
BOSSBS-3667R-A647
Anti-SLC44A1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antilichamen
Choline transporter. May be involved in membrane synthesis and myelin production.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLC44A1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR