Anti-ANO1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-3794R-FITC
BOSSBS-3794R-FITCEA 900 EUR
BOSSBS-3794R-FITC
Anti-ANO1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antilichamen
Calcium-activated chloride channel (CaCC) which plays a role in transepithelial anion transport and smooth muscle contraction. Required for the normal functioning of the interstitial cells of Cajal (ICCs) which generate electrical pacemaker activity in gastrointestinal smooth muscles. Acts as a major contributor to basal and stimulated chloride conductance in airway epithelial cells and plays an important role in tracheal cartilage development.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ANO1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR