Anti-UCHL1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-3806R-A555
BOSSBS-3806R-A555EA 901 EUR
BOSSBS-3806R-A555
Anti-UCHL1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antilichamen
Ubiquitin-protein hydrolase involved both in the processing of ubiquitin precursors and of ubiquitinated proteins. This enzyme is a thiol protease that recognizes and hydrolyzes a peptide bond at the C-terminal glycine of ubiquitin. Also binds to free monoubiquitin and may prevent its degradation in lysosomes. The homodimer may have ATP-independent ubiquitin ligase activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: UCHL1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Specificaties

antigeen symbool UCHL1
Type antilichaam Primair,Ubiquitin Carboxyl-Terminal Esterase L1
Environmentally Preferable
Antigeen synoniemen Neuron cytoplasmic protein 9.5|Ubiquitin thioesterase L1.|PGP9.5|Ubiquitin carboxyl-terminal|hydrolase isozyme L1|Ubiquitin thioesterase L1|UCH-L1|PGP 9.5|PARK5
Type antilichaam Primair,Ubiquitin Carboxyl-Terminal Esterase L1
Clonality Polyklonaal
Conjugatie Alexa Fluor® 555
Reactivity Human,Rat,Mouse
Host Rabbit
Gen-ID 7345
Isotype IgG
ImmunoChemie Yes
Immunogeen 30-80/223
Formulier liquid
Purification Purified by Protein A
Concentratie 1 μg/μl
Kruisadsorptie No
Opslagbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Opslagtemperatuur Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR