Anti-ADCY10 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-3916R-HRP
BOSSBS-3916R-HRPEA 900 EUR
BOSSBS-3916R-HRP
Anti-ADCY10 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antilichamen
Soluble adenylyl cyclase that has a critical role in mammalian spermatogenesis. Produces the cAMP which mediates in part the cAMP-responsive nuclear factors indispensable for maturation of sperm in the epididymis. Induces capacitation, the maturational process that sperm undergo prior to fertilization. May be the bicarbonate sensor. Involved in ciliary beat regulation.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: ADCY10
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR