Anti-COX1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Leverancier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-3953R-A555
BOSSBS-3953R-A555EA 901 EUR
BOSSBS-3953R-A555
Anti-COX1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antilichamen
Cytochrome c oxidase subunit I (COI or MTCO1) is one of three mitochondrial DNA (mtDNA) encoded subunits (MTCO1, MTCO2, MTCO3) of respiratory Complex IV. Complex IV is located within the mitochondrial inner membrane and is the third and final enzyme of the electron transport chain of mitochondrial oxidative phosphorylation. Complex IV is composed of 13 polypeptides. Subunits I, II, and III (MTCO1, MTCO2, MTCO3) are encoded by mtDNA while subunits IV, Va, Vb, VIa, VIb, VIc, VIIa, VIIb, VIIc, and VIII are nuclear encoded. Mammalian MTCO1 has 12 membrane-spanning alpha-helices (I to XII).