Anti-DEGS1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-4057R-A750
BOSSBS-4057R-A750EA 890 EUR
BOSSBS-4057R-A750
Anti-DEGS1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antilichamen
DEGS1 is a member of the membrane fatty acid desaturase family which is responsible for inserting double bonds into specific positions in fatty acids. It contains three His containing consensus motifs that are characteristic of a group of membrane fatty acid desaturases. It has sphingolipid-delta-4-desaturase activity and converts D-erythro-sphinganine to D-erythro-sphingosine (E-sphing-4-enine).

Type: Primary
Antigen: DEGS1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 21-120/323
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Specificaties

Environmentally Preferable
Type antilichaam Primair,DEGS1
Antigeen synoniemen DEGS 1|Membrane fatty acid lipid desaturase|Sphingolipid delta4 desaturase DES1.DEGS1_HUMAN|Des 1|Degenerative spermatocyte homolog lipid desaturase|Degenerative spermatocyte homolog 1|Dihydroceramide desaturase|Degenerative spermatocyte homolog 1 lipid desaturase|MIG15v Migration inducing gene 15 protein|MLD|MGC5079|Sphingolipid delta 4 desaturase|FADS7|Membrane lipid desaturase|DES1|DEGS
Type antilichaam Primair,DEGS1
Clonality Polyklonaal
Conjugatie Alexa Fluor® 750
Reactivity
Host Rabbit
Gen-ID 653121
Isotype IgG
ImmunoChemie Yes
ImmunoFluorescentie Yes
Modification Unmodified
Epitoop 21-120/323
Immunogeen KLH conjugated synthetic peptide derived from human DEGS1
Formulier Liquid
Moleculair gewicht 35 kDa
Purification Purified by Protein A
Concentratie 1 µg/µl
Opslagbuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Opslagtemperatuur Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Shipping temperature 4 °C
Tested applications ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR