Anti-CARM1 Rabbit Polyclonal Antibody (Cy7®)
Leverancier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-4098R-CY7
BOSSBS-4098R-CY7EA 900 EUR
BOSSBS-4098R-CY7
Anti-CARM1 Rabbit Polyclonal Antibody (Cy7®)
Antilichamen
Methylates (mono- and asymmetric dimethylation) the guanidino nitrogens of arginyl residues in several proteins involved in DNA packaging, transcription regulation, pre-mRNA splicing, and mRNA stability. Recruited to promoters upon gene activation together with histone acetyltransferases from EP300/P300 and p160 families, methylates histone H3 at 'Arg-17' (H3R17me), forming mainly asymmetric dimethylarginine (H3R17me2a), leading to activate transcription via chromatin remodeling. During nuclear hormone receptor activation and TCF7L2/TCF4 activation, acts synergically with EP300/P300 andMore Product Information