Anti-VLDL Receptor Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-4228R-A750
BOSSBS-4228R-A750EA 899 EUR
BOSSBS-4228R-A750
Anti-VLDL Receptor Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antilichamen
Binds VLDL and transports it into cells by endocytosis. In order to be internalized, the receptor-ligand complexes must first cluster into clathrin-coated pits. Binding to Reelin induces tyrosine phosphorylation of Dab1 and modulation of Tau phosphorylation (By similarity).

Type: Primary
Antigen: VLDL Receptor
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 551-650/873
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR