Anti-ADORA1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-4235R-A647
BOSSBS-4235R-A647EA 893 EUR
BOSSBS-4235R-A647
Anti-ADORA1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antilichamen
Receptor for adenosine. The activity of this receptor is mediated by G proteins which inhibit adenylyl cyclase.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADORA1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Specificaties

antigeen symbool ADORA1
Environmentally Preferable
Type antilichaam Adenosine A1 Receptor,Primair
Antigeen synoniemen RDC7
Type antilichaam Adenosine A1 Receptor,Primair
Clonality Polyklonaal
Conjugatie Alexa Fluor® 647
Reactivity Human,Rat,Mouse
Host Rabbit
Gen-ID 134
Isotype IgG
ImmunoChemie Yes
Immunogeen 147-176/326
Formulier liquid
Purification Purified by Protein A
Concentratie 1 μg/μl
Kruisadsorptie No
Opslagbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Opslagtemperatuur Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR