Anti-COL4A3BP Rabbit Polyclonal Antibody (Cy5.5®)

Leverancier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-5044R-CY5.5
BOSSBS-5044R-CY5.5EA 900 EUR
BOSSBS-5044R-CY5.5
Anti-COL4A3BP Rabbit Polyclonal Antibody (Cy5.5®)
Antilichamen
Shelters ceramides and diacylglycerol lipids inside its START domain and mediates the intracellular trafficking of ceramides and diacylglycerol lipids in a non-vesicular manner.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: COL4A3BP
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR