Anti-CHEK2 Rabbit Polyclonal Antibody (Cy5.5®)
Leverancier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-5275R-CY5.5
BOSSBS-5275R-CY5.5EA 930 EUR
BOSSBS-5275R-CY5.5
Anti-CHEK2 Rabbit Polyclonal Antibody (Cy5.5®)
Antilichamen
Chk2 is a serine/threonine kinase involved in the control of cell cycle checkpoints, and may also participate in transduction of the DNA damage and replicational stress signals. Chk2 is the mammalian ortholog of the budding yeast Rad53 and fission yeast Cds1 checkpoint kinases. The amino-terminal domain of Chk2 contains a series of seven serine and threonine residues (Ser19, Thr26, Ser28, Ser33, Ser35, Ser50 and Thr68) followed by glutamine (SQ or TQ motif). These are known to be preferred sites for phosphorylation by ATM/ATR kinases. Indeed, after DNA damage by ionizing radiation (IR), UVMore Product Information