Anti-GRIA2 Rabbit Polyclonal Antibody (Cy5.5®)
Leverancier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-5359R-CY5.5
BOSSBS-5359R-CY5.5EA 930 EUR
BOSSBS-5359R-CY5.5
Anti-GRIA2 Rabbit Polyclonal Antibody (Cy5.5®)
Antilichamen
Glutamate receptors are the predominant excitatory neurotransmitter receptors in the mammalian brain and are activated in a variety of normal neurophysiologic processes. This gene product belongs to a family of glutamate receptors that are sensitive to alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate(AMPA), and function as ligand-activated cation channels. These channels are assembled from 4 related subunits, Gria1-4. The subunit encoded by this gene (Gria2) is subject to RNA editing(CAG->CGG; Q->R) within the second transmembrane domain, which is thought to render the channelMore Product Information